Anti-GZMA Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-2578R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Granzyme A
- Antigen symbol:GZMA
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3001
- Antigen synonyms:Ganzyme 1 cytotoxic T-lymphocyte-associated serine esterase 3,Ctla-3|SE1|TSP-1|Hf|Ctla3|TSP1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:110-160/262
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Abundant protease in the cytosolic granules of cytotoxic T-cells and NK-cells which activates caspase-independent cell death with morphological features of apoptosis when delivered into the target cell through the immunological synapse. It cleaves after Lys or Arg. Cleaves APEX1 after 'Lys-31' and destroys its oxidative repair activity. Cleaves the nucleosome assembly protein SET after 'Lys-189', which disrupts its nucleosome assembly activity and allows the SET complex to translocate into the nucleus to nick and degrade the DNA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GZMA/Granzyme A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat