Order Entry
Export
ContactUsLinkComponent
Anti-HTR4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-HTR4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-2127R-A488
Supplier:  Bioss
Anti-HTR4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-2127R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5-Hydroxytryptamine (Serotonin) Receptor 4, G Protein-Coupled
  • Antigen symbol:
    HTR4
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    3360
  • Antigen synonyms:
    5-HT4R|5-HT4
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    41-58/388
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that stimulate adenylate cyclase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HTR4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat