Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 22
- Antigen symbol:CCL22
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:6367
- Antigen synonyms:ABCD-1|SCYA22|MDC|A-152E5.1|DC/B-CK|STCP-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The MDC gene is one of several Cys-Cys (CC) cytokine genes which are clustered on the q arm of chromosome 16. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by MDC displays chemotactic activity for natural killer cells, chronically activated T lymphocytes, monocytes and dendritic cells. It has no chemoattractant activity for eosinophils, neutrophils and resting T lymphocytes and also displays a mild activity for primary activated T lymphocytes. The product of this gene binds to chemokine receptor CCR4. This chemokine may play a role in the trafficking of activated T lymphocytes to inflammatory sites and other aspects of activated T lymphocyte physiology.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CCL22
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat