Anti-GAP43 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-1641R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Growth Associated Protein 43
- Antigen symbol:GAP43
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2596
- Antigen synonyms:axonal membrane protein GAP-43|growth-associated protein 43|neuron growth-associated protein 43 protein F1|neuromodulin|nerve growth-related peptide GAP43|calmodulin-binding protein P-57|neural phosphoprotein B-50|PP46|B-50
- Modification:phospho-Ser41
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The protein encoded by this gene has been termed a 'growth' or 'plasticity' protein because it is expressed at high levels in neuronal growth cones during development and axonal regeneration. This protein is considered a crucial component of an effective regenerative response in the nervous system. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq, Jul 2008]
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GAP43
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat