Anti-GEN1 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-13334R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:GEN1 Holliday junction 5' flap endonuclease
- Antigen symbol:GEN1
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Gen
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Endonuclease which cleaves flap structures at the junction between single-stranded DNA and double-stranded DNA. Specific for 5'-overhanging flap structures in which the 5'-upstream of the flap is completely double-stranded. Prefers the blocked-flap structures similar to those occurring at replication forks, in which the 5' single-strand overhang of the flap is double-stranded.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GEN1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat