Specifications
- Antibody type:Primary
- Antigen name:Kelch-like 28
- Antigen symbol:KLHL28
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:54813
- Antigen synonyms:BTBD5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
KLHL28 is a 571 amino acid protein similar to the Drosophila kelch protein. KLHL26 contains six kelch repeats and one BTB (POZ) domain. The BTB (broad complex, tramtrack and bric-a-brac) domain, also known as the POZ (Poxvirus and zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. KLHL28 is expressed as two isoforms produced by alternative splicing.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KLHL28
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat