Order Entry
Export
ContactUsLinkComponent
Anti-KLHL28 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-KLHL28 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-12907R-A555
Supplier:  Bioss
Anti-KLHL28 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-12907R-A555
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Kelch-like 28
  • Antigen symbol:
    KLHL28
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    54813
  • Antigen synonyms:
    BTBD5
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

BTBD14A is a 587 amino acid protein that contains one BTB/POZ domain. The BTB/POZ domain mediates homomeric and heteromeric POZ-POZ interactions and is common to transcriptional regulators involved in chromatin modeling. In several BTB/POZ containing proteins, including BCL-6 and the promyelocytic leukemia zinc-finger (PLZF) oncoprotein, this domain interacts with the SMRT/N-CoR-mSin3A HDAC complex and is directly involved in repressing and silencing gene transcription. When this domain is deleted, as with the oncogenic PLZF-RAR chimera of promyelocytic leukemias, this transcriptional repression is attenuated. This suggests that BTBD14A may play a role in transcription regulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLHL28
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat