Anti-CHRM3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-1289R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Cholinergic receptor, muscarinic 3
- Antigen symbol:CHRM3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:12671
- Antigen synonyms:HM3|PBS|EGBRS
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:546-589/589
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The muscarinic acetylcholine receptor mediates various cellular responses, including inhibition of adenylate cyclase, breakdown of phosphoinositides and modulation of potassium channels through the action of G proteins. Primary transducing effect is Pi turnover.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CHRM3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity: