Specifications
- Antibody type:Primary
- Antigen name:Fucosyltransferase 11
- Antigen symbol:FUT11
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:170384
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Glycosyltransferases that mediate the regio- and stereoselective transfer of sugars, such as the fucosyltransferases, determine cell surface-carbohydrate profiles, which are essential interfaces for biological recognition processes. Fucosyltransferases (FucTs) catalyze the covalent association of fucose to different positional linkages on sugar acceptor molecules. The carbohydrate moieties that are generated are covalently attached to cell surfaces and are necessary to ensure a surface contour that satisfies a variety of physiological roles. FucT-XI is a 492 amino acid single-pass type II membrane protein that belongs to the glycosyltransferase 10 family. Localizing to Golgi apparatus, FucT-XI may act as a fucosyltransferase and exists as two alternatively spliced isoforms. The gene encoding FucT-XI maps to mouse chromosome 14 A3.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FUT11/FucT-XI
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat