Order Entry
Export
ContactUsLinkComponent
 
Anti-ZNF750 Rabbit Polyclonal Antibody (Cy5®)
  BOSSBS-12239R-CY5
 :  Bioss
Anti-ZNF750 Rabbit Polyclonal Antibody (Cy5®)
  BOSSBS-12239R-CY5
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Zinc Finger Protein 750
  • Antigen symbol:
    ZNF750
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    79755
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Zinc finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc finger proteins contain a Krüppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. Zinc finger protein 750 is a 723 amino acid member of the Krüppel C2H2-type zinc finger protein family. Localized to the nucleus, ZNF750 contains one conserved C2H2 zinc finger domain and is expressed in the skin, lungs, prostate, placenta and thymus. ZNF750 is also expressed in primary human keratinocytes but not in fibroblasts. Mutations in the gene encoding ZNF750 cause Seborrhea-like dermatitis with psoriasiform, a condition characterized by a chronic and diffuse rash on the face and hyperkeratosis of skin over the elbows, soles, knees and palms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF750
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat