Order Entry
Export
ContactUsLinkComponent
Anti-FAM62C Rabbit Polyclonal Antibody (Cy7®)
Anti-FAM62C Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-12165R-CY7
 :  Bioss
Anti-FAM62C Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-12165R-CY7
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Family With Sequence Similarity 62, Member C
  • Antigen symbol:
    FAM62C
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    ESYT3|E-Syt3|FAM62C|ESYT3_HUMAN.|Extended synaptotagmin-like protein 3|Extended synaptotagmin 3|CHR3SYT
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Chromosome 3 is made up of about 214 million bases encoding over 1,100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumor suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesis suppressor SEMA3B. Marfan Syndrome, porphyria, von Hippel-Lindau syndrome, osteogenesis imperfecta and Charcot-Marie-Tooth Disease are a few of the numerous genetic diseases associated with chromosome 3. The FAM62C gene product has been provisionally designated FAM62C pending further characterization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM62C
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat