Specifications
- Antibody type:Primary
- Antigen name:G protein coupled receptor 33
- Antigen symbol:GPCR33
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:gprc6a|GPCR 6A|hGPCR33|GPR C6A|G protein coupled receptor 33|G-protein coupled receptor GPCR33|hGPCR 33|GPCR|GPC6A_HUMAN|hGPRC6A|G-protein coupled receptor family C group 6 member A|GPRC 6A|GPCR 33|bA86F4.3|OTTHUMP00000017075.|GPCR33|G protein coupled receptor family C group 6 member A
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
GPRC6A (G protein-coupled receptor family C group 6 member A) is a 928 amino acid protein encoded by the mouse Gprc6a gene. GPRC6A is a member of the G protein-coupled receptor 3 family. This receptor is activated by both amino acids and extracellular concentration of calcium ions. The activity of GPRC6A is mediated by a G protein that activates a phosphatidylinositol-calcium second messenger system. GPRC6A senses changes in the extracellular concentration of calcium ions, suggesting that it may mediate extracellular calcium-sensing responses in osteoblasts. Osteocalin stimulates the activity of GPRC6A in the presence of calcium. GPRC6A also acts as a receptor for amino acids, with a preference for basic amino acids such as L-Lys, L-Arg and L-ornithine. Its affinity for amino acids suggests that it may act as a regulatory component of the urea cycle. GPRC6A is expressed at high levels in liver, lung, spleen and heart, and at lower levels in kidney, skeletal muscle and brain.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: GPCR6A/GPCR33
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat