Order Entry
Export
ContactUsLinkComponent
Anti-SLC25A12 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-SLC25A12 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-11953R-HRP
Supplier:  Bioss
Anti-SLC25A12 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-11953R-HRP
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 25 Member A12
  • Antigen symbol:
    SLC25A12
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    solute carrier family 25|ARALAR1|CMC1_HUMAN|Araceli hiperlarga|Mitochondrial aspartate glutamate carrier 1|Calcium-binding mitochondrial carrier protein Aralar1|Solute carrier family 25 member 12|member 12.|ARALAR|Calcium binding mitochondrial carrier superfamily member|AGC1|SLC25A12
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Calcium signaling in mitochondria is important in order for it to function in response to a variety of extracellular stimuli. Signaling begins with Ca(2+) entry in mitochondria via the Ca(2+) uniporter followed by Ca(2+) activation of three dehydrogenases in the mitochondrial matrix. ARALAR, the neuronal Ca(2+)-binding mitochondrial aspartate-glutamate carrier, has Ca(2+) binding domains facing the extramitochondrial space and functions in the malate-aspartate NADH shuttle (MAS). ARALAR is encoded by the SLC25A12 gene and is expressed in brain and skeletal muscle. ARALAR is required for the synthesis of brain aspartate and N-acetylaspartatemay and plays a role in myelin formation. It is also essential for the transmission of small Ca(2+) signals to mitochondria via an increase in mitochondrial NADH. In addition, ARALAR is implicated in conferring susceptibility to schizophrenia.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SLC25A12
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat