Anti-MAOB Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-11297R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Monoamine Oxidase B
- Antigen symbol:MAOB
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:4129
- Antigen synonyms:HGNC:6834|monoamine oxidase B |MAOB|platelet|RP1-201D17__B.1|MAO|brain|adrenalin oxidase|amine oxidase (flavin-containing)|MGC26382|tyramine oxidase
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:50-100/520
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Catalyzes the oxidative deamination of biogenic and xenobiotic amines and has important functions in the metabolism of neuroactive and vasoactive amines in the central nervous system and peripheral tissues. MAOB preferentially degrades benzylamine and phenylethylamine.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MAOB
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat