Anti-SRC Rabbit Polyclonal Antibody (Cy3®)
Product Details & Documents
c-Src tyrosine kinase plays a critical role in signal transduction downstream of growth factor receptors, integrins and G protein-coupled receptors. We used stable isotope labeling with amino acids in cell culture (SILAC) approach to identify additional substrates of c-Src tyrosine kinase in human embryonic kidney 293T cells. We have identified 10 known substrates and interactors of c-Src and Src family kinases along with 26 novel substrates. We have experimentally validated 4 of the novel proteins (NICE-4, RNA binding motif 10, FUSE-binding protein 1 and TRK-fused gene) as direct substrates of c-Src using in vitro kinase assays and cotransfection experiments. Significantly, using a c-Src specific inhibitor, we were also able to implicate 3 novel substrates (RNA binding motif 10, EWS1 and Bcl-2 associated transcription factor) in PDGF signaling. Finally, to identify the exact tyrosine residues that are phosphorylated by c-Src on the novel c-Src substrates, we designed custom peptide microarrays containing all possible tyrosine-containing peptides (312 unique peptides) and their mutant counterparts containing a Tyr -->Phe substitution from 14 of the identified substrates. Using this platform, we identified 34 peptides that are phosphorylated by c-Src. We have demonstrated that SILAC-based quantitative proteomics approach is suitable for identification of substrates of nonreceptor tyrosine kinases and can be coupled with peptide microarrays for high-throughput identification of substrate phosphopeptides.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SRC
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
bs-1135R-CY3
Datasheet
Specifications
- Clonality:Polyclonal
- Purification:Purified by Protein A
- Antibody type:Primary
- Form:liquid
- Isotype:IgG
- Storage temperature:Store at 4°C for 12 months
- Host:Rabbit
- Concentration:1 μg/μl
- Cross adsorption:No
- Shipping temperature:4°C
- Antigen name:Proto-oncogene tyrosine-protein kinase Src
- Cat. No.:BOSSBS-1135R-CY3
- Reactivity:Human|Rat|Mouse
- Gene ID:6714
- Antigen synonyms:ASV|p60-Src|c-SRC|Steroid Receptor Coactivator|SRC1
- ImmunoChemistry:Yes
- Conjugation:Cy3®
- Antigen symbol:SRC
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.



