Specifications
- Antibody type:Primary
- Antigen symbol:HSP 40-4/HDJ2/DNAJA1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
DnaJ-like proteins interact with HSP 70 molecular chaperones and function to facilitate protein folding and mitochondrial protein import. HSP 40-4, also known as HDJ2, is the human DnaJ homolog that functions as a co-chaperone with a cysteine-rich zinc finger domain. The cellular redox enzyme thioredoxin interacts with HSP 40-4, and oxidation and reduction reversibly regulate HSP 40-4 function in response to the changing redox states of the cell. The zinc finger domain of HSP 40-4 may act as a redox sensor of chaperone-mediated protein-folding machinery, since HSP 40-4 inactivation leads to the oxidation of cysteine thiols and a simultaneous release of coordinated zinc. Loss of the HSP 40-4 protein may be linked to severe defects in spermatogenesis that involve aberrant androgen signaling.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HSP 40-4/HDJ2/DNAJA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat