Anti-FLOT2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-11254R-A488
: Bioss
- Antibody type:Primary
- Antigen name:Flotillin 2
- Antigen symbol:FLOT2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2319
- Antigen synonyms:ECS-1|membrane component|ECS1|FLOT2|flotillin 2|reggie-1|surface marker 1 (35kD protein identified by monoclonal ECS-1)|chromosome 17|ESA|M17S1|Flotillin 2 (epidermal surface antigen 1)|ESA1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:180-230/428
- Purification:Purified by Protein A
- Pk:100 µl
May act as a scaffolding protein within caveolar membranes, functionally participating in formation of caveolae or caveolae-like vesicles. May be involved in epidermal cell adhesion and epidermal structure and function.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ECS1/Flotillin 2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat