Anti-ARHGAP26 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-11248R-A647
: Bioss
- Antibody type:Primary
- Antigen name:Rho GTPase Activating Protein 26
- Antigen symbol:ARHGAP26
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:GRAF|OPHN1L1|OPHN1L|GRAF1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Cellular signaling by G-proteins is down-regulated by GTPase-activating proteins (GAPs), which increase the rate of GTP hydroylsis. The GTPase regulator associated with focal adhesion kinase (GRAF) has GAP activity toward Rho A and Cdc42, but not Rac1. GRAF is ubiquitously expressed with high levels in heart and brain. Expression of GRAF causes clearing of stress fibers and formation of long actin based filopodial-like extensions. Fusion of MLL with GRAF, MLL/GRAF, is included in a rare genetic subgroup of acute myeloid leukemia (AML) cases.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ARHGAP26
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat