Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger And BTB Domain-Containing protein 17
- Antigen symbol:ZBTB17
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Myc-interacting zinc finger protein 1|Zinc finger and BTB domain containing protein 17|Zinc finger protein 60|Myc-interacting zinc finger protein|Zinc finger and BTB domain-containing protein 17|Miz-1|ZNF60.|ZNF151|ZBT17_HUMAN|Zbtb17|Zinc finger protein 151|ZBT17
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The Myc family, including c-Myc-, N-Myc- and L-Myc, are nuclear proteins with relatively short half lives that contribute an important role in cellular processes such as proliferation, differentiation, apoptosis and transformation. The c-Myc protein activates transcription as part of a heteromeric complex with a number of interacting partners, including Max and Mxi 1; however the transforming properties of the Myc proto-oncogene are believed to be associated with Myc-mediated transcriptional repression. A POZ domain Zn finger protein, designated Miz-1 for Myc-interacting Zn finger protein-1, is a specific target of Myc-induced gene repression. Miz-1 interacts with Myc, but not Max or other Myc partners, and binding of Myc to Miz-1 requires the helix-loop-helix domain of Myc and a short amphipathic helix located in the carboxy-terminus of Miz-1. Miz-1 associates with DNA elements on the adenovirus major late and cyclin D1 promoters and activates transcription of both promoters. Expression of Miz-1 induces potent growth arrest function, and this latency is reversed by the addition of Myc.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ZBTB17
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat