Specifications
- Antibody type:Primary
- Antigen name:Proto-Cadherin gamma subfamily C4
- Antigen symbol:PCDHGC4
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters, designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. PCDHGC4 (protocadherin gamma subfamily C, 4) is a 938 amino acid single-pass type I membrane protein that localizes to the cell membrane and contains six cadherin domains. Expressed as two alternatively spliced isoforms, PCDHGC4 functions as a potential calcium-dependent cell adhesion protein that is thought to be involved in the establishment and maintenance of neuronal connections within the brain. The gene encoding PCDHGC4 maps to a protocadherin gamma gene cluster which is localized to chromosome 5 and contains over 22 protocadherin genes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PCDHGC4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat