Anti-NPY1R Rabbit Polyclonal Antibody (Cy5®)
BOSSBS-1070R-CY5
: Bioss
- Antibody type:Primary
- Antigen name:Neuropeptide Y Receptor Y1
- Antigen symbol:NPY1R
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:4886
- Antigen synonyms:NPY1-R|NPYR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:330-382/382
- Purification:Purified by Protein A
- Pk:100 µl
Receptor for neuropeptide Y and peptide YY. The rank order of affinity of this receptor for pancreatic polypeptides is NPY >[Pro-34] PYY, PYY and [Leu-31, Pro-34] NPY >NPY (2-36) >[Ile-31, Gln-34] PP and PYY (3-36) >PP >NPY free acid.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NPY1R
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat