Anti-ZNF268 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
BOSSBS-0626R-HRP
: Bioss
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 268
- Antigen symbol:ZNF268
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:138-149/178
- Purification:Purified by Protein A
- Pk:100 µl
Recommended Dilutions: IHC-P: 1:100-500
Type: Primary
Antigen: ZNF268
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human