Specifications
- Antibody type:Primary
- Antigen name:Cyclin E1
- Antigen symbol:CCNE1
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:25729
- Antigen synonyms:G1/S-specific cyclin-E1|cyclin Et|cyclin Es|CCNE|CYCLE,CCNEcyclin Es
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:370-411/411
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The protein encoded by this gene belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. This cyclin forms a complex with and functions as a regulatory subunit of CDK2, whose activity is required for cell cycle G1/S transition. This protein accumulates at the G1-S phase boundary and is degraded as cells progress through S phase. Overexpression of this gene has been observed in many tumors, which results in chromosome instability, and thus may contribute to tumorigenesis. This protein was found to associate with, and be involved in, the phosphorylation of NPAT protein (nuclear protein mapped to the ATM locus), which participates in cell-cycle regulated histone gene expression and plays a critical role in promoting cell-cycle progression in the absence of pRB. Two alternatively spliced transcript variants of this gene, which encode distinct isoforms, have been described. Two additional splice variants were reported but detailed nucleotide sequence information is not yet available. Transcript Variant: This variant (1) contains a different 5' end region, which includes an upstream in-frame translation start codon, when compared to variant 2. The encoded protein has a 15 aa longer N-terminus, as compared to isoform 2.
Recommended Dilutions: Flow Cytometry: 1:20-100
Type: Primary
Antigen: CCNE1
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat