Anti-GAP43 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-0154R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Growth Associated Protein 43
- Antigen symbol:GAP43
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:14432
- Antigen synonyms:axonal membrane protein GAP-43|growth-associated protein 43|neuron growth-associated protein 43 protein F1|neuromodulin|nerve growth-related peptide GAP43|calmodulin-binding protein P-57|neural phosphoprotein B-50|PP46|B-50
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:1-50/227
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This protein is associated with nerve growth. It is a major component of the motile "growth cones" that form the tips of elongating axons. Plays a role in axonal and dendritic filopodia induction.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GAP43
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat