Order Entry
Export
ContactUsLinkComponent
Anti-CASP4 + CASP 13 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-CASP4 + CASP 13 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-0053R-HRP
Supplier:  Bioss
Anti-CASP4 + CASP 13 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BOSSBS-0053R-HRP
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Caspase 4 And Caspase 13
  • Antigen symbol:
    CASP4 + CASP 13
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Bovine
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    Caspase 13 subunit 2|CASP-13|apoptosis related cysteine|Caspase-13|ERICE.|CASPD_BOVIN|Caspase 4|Caspase 13 subunit 1|CASP4|Caspase 4 subunit 1|Evolutionary related interleukin-1-beta-converting enzyme
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Caspase-13 (also designated Evolutionarily related interleukin-1beta converting enzyme ; ERICE) involved in the activation cascade of caspases responsible for apoptosis execution. Might function by either activating some proteins required for cell death or inactivating proteins necessary for cell survival. [subunit] Heterodimer of a small and a large subunit. The two subunits are derived from the precursor sequence by an autocatalytic mechanism or by cleavage by Caspase-8. Mainly expressed in peripheral blood lymphocytes, spleen and placenta. Belongs to peptidase family C14.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CASP4 + CASP 13
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Bovine