Order Entry
Export
ContactUsLinkComponent
Anti-Gap-43 Rabbit Polyclonal Antibody
Anti-Gap-43 Rabbit Polyclonal Antibody
Catalog # BSENR-1651-100
Supplier:  Biosensis
undefined
Anti-Gap-43 Rabbit Polyclonal Antibody
Catalog # BSENR-1651-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neuromodulin
  • Antigen symbol:
    GAP43
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal peptide 217-227 of rat and mouse GAP43, which is KEDPEADQEHA, to which an N terminal Cysteine residue was added to allow chemical coupling to Keyhole Limpet Hemocyanin carrier protein.
  • Format:
    Affinity purified
  • Form:
    Lyophilized
  • Gene ID:
    P06837 NEUM_MOUSE
  • Antigen synonyms:
    Axonal membrane protein GAP-43|Growth-associated protein 43|Calmodulin-binding protein P-57
  • Storage buffer:
    PBS, pH 7.4, with 3% Trehalose, without preservatives
  • Sequence:
    KEDPEADQEHA
  • Storage temperature:
    Store lyophilised, unopened vial at 2-8 °C or lower. After reconstitution, prepare aliquots and store at −20 °C to −80 °C for a higher stability.
  • Shipping temperature:
    Room (ambient) temperature.
  • Immunogen:
    C-terminal peptide 217-227 of rat and mouse GAP43, which is KEDPEADQEHA, to which an N terminal Cysteine residue was added to allow chemical coupling to Keyhole Limpet Hemocyanin carrier protein.
  • Purification:
    Affinity purified.
  • Size:
    100 ug
  • Pk:
    100 µG

Specifications

About this item

GAP43 is very abundant protein which is found concentrated in neurons. One group discovered it as one of three proteins which becomes unregulated during the regeneration of the toad optic nerve (1). Three GAPs (Growth associated proteins) were discovered, and the number 43 comes from the apparent SDS-PAGE molecular weight of the one named GAP43. The HGNC name for this protein is, not surprisingly, GAP43. Later work showed that GAP43 does not run on SDS-PAGE in a fashion which accurately reflects its molecular weight, and that GAP43 proteins from different species may run at different apparent molecular weights. Partly due to these features GAP43 were independently discovered by several different groups and therefore has several alternate names, such as protein F1, pp46, neuromodulin, neural phosphoprotein B-50 and calmodulin-binding protein P-57. In each case the number reflects the apparent SDS-PAGE molecular weight, and underlines the unusual properties of this molecule. Mammalian GAP43 proteins contains only 226-243 amino acids, and so the real molecular weight is 23.61-25.14 kDa. GAP43 has been extensively studied and is known to be a major protein kinase C substrate and to bind calmodulin avidly. GAP43 is anchored to the plasma membrane by palmitoylation modifications.

Western Blotting (WB). A dilution of 1:5,000 - 1:20,000 is recommended. A dilution of 1:500-2,000 is recommended for ICC/IHC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Gap-43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal peptide 217-227 of rat and mouse GAP43, which is KEDPEADQEHA, to which an N terminal Cysteine residue was added to allow chemical coupling to Keyhole Limpet Hemocyanin carrier protein.
Host: Rabbit
Isotype:
Reactivity: Human;Rat;Mouse;Bovine