Chloridazon standard
EHERC11320000
:
- Concentration:250 mg
- Pk:0,25 g
- CAS No.:1698-60-8
- Description:Chloridazon
- Formula:C₁₀H₈ClN₃O
- Description:Chloridazon
Synonym: 5-Amino-4-chloro-2-methylpyridazin-3-one
Synonym: 5-Amino-4-chloro-2-methylpyridazin-3-one