Anti-MMP-14 Rabbit Polyclonal Antibody [clone: P509]
BWRLBS1232
- Antibody type:Primary
- Antigen symbol:MMP-14
- Clonality:Polyclonal
- Clone:P509
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 480-520 of Human MMP-14.
MMP-14 (P509) pAb detects endogenous levels of MMP-14 protein.
Type: Primary
Antigen: MMP-14
Clonality: Polyclonal
Clone: P509
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat