Order Entry
Export
ContactUsLinkComponent
 
Anti-Chk2 Rabbit Polyclonal Antibody [clone: S62]
  BWRLBS1055
Anti-Chk2 Rabbit Polyclonal Antibody [clone: S62]
  BWRLBS1055

 

  • Antibody type:
    Primary
  • Antigen name:
    Checkpoint Kinase 2
  • Antigen symbol:
    CHK2
  • Clonality:
    Polyclonal
  • Clone:
    S62
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 31-80 of Human Chk2.

Chk2 (S62) pAb detects endogenous levels of Chk2 protein.

Type: Primary
Antigen: Chk2
Clonality: Polyclonal
Clone: S62
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat