Order Entry
Export
ContactUsLinkComponent
Anti-Cleaved-PARP-1 Rabbit Polyclonal Antibody [clone: D214]
Anti-Cleaved-PARP-1 Rabbit Polyclonal Antibody [clone: D214]
Catalog # BWRLBS7047
Anti-Cleaved-PARP-1 Rabbit Polyclonal Antibody [clone: D214]
Catalog # BWRLBS7047

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Cleaved-PARP-1
  • Clonality:
    Polyclonal
  • Clone:
    D214
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 180-214 of Human PARP-1.

Cleaved-PARP-1 (D214) pAb detects endogenous levels of the small fragment (24 kDa) of PARP1 resulting from caspase cleavage.

Type: Primary
Antigen: Cleaved-PARP-1
Clonality: Polyclonal
Clone: D214
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat