Anti-Cleaved-MMP-15 Rabbit Polyclonal Antibody [clone: Y132]
BWRLBS7041
- Antibody type:Primary
- Antigen symbol:Cleaved-MMP-15
- Clonality:Polyclonal
- Clone:Y132
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 100-150 of Human MMP-15.
Cleaved-MMP-15 (Y132) pAb detects endogenous levels of Cleaved-MMP-15 (60 kDa) protein.
Type: Primary
Antigen: Cleaved-MMP-15
Clonality: Polyclonal
Clone: Y132
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat