Anti-ECE1 Rabbit Polyclonal Antibody
100 µg
Catalog # BWRLBS7649
Supplier: Bioworld Technology
Jump to:
Product Details & Documents
Recombinant full length Human ECE1.
ECE1 pAb detects endogenous levels of ECE1 protein.
Type: Primary
Antigen: ECE1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Specifications
- Size:100 µg
- Western blot:Yes
- Cat. No.:BWRLBS7649
- Clonality:Polyclonal
- Reactivity:Human|Rat|Mouse
- Antibody type:Primary
- Host:Rabbit
- ImmunoChemistry:Yes
- Antigen symbol:ECE1



