Anti-Catenin-alpha1/2 Rabbit Polyclonal Antibody
BWRLBS6976
- Antibody type:Primary
- Antigen symbol:Catenin-alpha1/2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human Catenin-alpha1.
Catenin-?1/2 pAb detects endogenous levels of Catenin-?1/2 protein.
Type: Primary
Antigen: Catenin-alpha1/2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat