Order Entry
Export
ContactUsLinkComponent
Anti-14-3-3 beta Rabbit Polyclonal Antibody
Anti-14-3-3 beta Rabbit Polyclonal Antibody
Catalog # BWRLBS6955
Anti-14-3-3 beta Rabbit Polyclonal Antibody
Catalog # BWRLBS6955

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    14-3-3 beta
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recombinant full length Human 14-3-3 beta.

14-3-3 ? pAb detects endogenous levels of 14-3-3 ? protein.

Type: Primary
Antigen: 14-3-3 beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat