Anti-HA-Tag Goat Antibody
BWRLBS6941
- Antibody type:Primary
- Antigen symbol:HA-Tag
- Host:Goat
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide HA-tag.
HA-tag pAb detects over-expressed or recombinant proteins containing the HA epitope tag.
Type: Primary
Antigen: HA-Tag
Clonality:
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human; Mouse; Rat