Anti-NFkB-p105/p50 Mouse Monoclonal Antibody
Catalog # BWRLMB0155
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:NFkB-p105/p50
- Clonality:Monoclonal
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human NFkB-p105.
This antibody detects endogenous levels of NFkB-p105/p50 and does not cross-react with related proteins
Type: Primary
Antigen: NFkB-p105/p50
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human