Order Entry
Export
ContactUsLinkComponent
Anti-Csk Mouse Monoclonal Antibody
Anti-Csk Mouse Monoclonal Antibody
  BWRLMB0131
Anti-Csk Mouse Monoclonal Antibody
  BWRLMB0131

 

  • Antibody type:
    Primary
  • Antigen symbol:
    Csk
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recombinant full length Human Csk.

This antibody detects endogenous levels of Csk and does not cross-react with related proteins.

Type: Primary
Antigen: Csk
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Rat