Anti-MASP2 Rabbit Polyclonal Antibody
BWRLBS5789
- Antibody type:Primary
- Antigen symbol:MASP2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 268-313 of Human MASP2.
MASP2 pAb detects endogenous levels of MASP2 protein.
Type: Primary
Antigen: MASP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse