Anti-p-IKKgamma Rabbit Polyclonal Antibody [clone: S31 ]
Catalog # BWRLBS4238
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-IKK gamma
- Clonality:Polyclonal
- Clone:S31
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human IKKgamma around the phosphorylation site of Serine 31.
p-IKK? (S31) pAb detects endogenous levels of IKK? protein only when phosphorylated at Ser31.
Type: Primary
Antigen: p-IKKgamma
Clonality: Polyclonal
Clone: S31
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat