Anti-SCT Rabbit Polyclonal Antibody
Catalog # BWRLBS60409
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:SCT
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
A synthetic peptide corresponding to residues in Human SCT.
SCT pAb detects endogenous levels of SCT protein.
Type: Primary
Antigen: SCT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat