Order Entry
Export
ContactUsLinkComponent
Anti-AMD1 Rabbit Polyclonal Antibody
Anti-AMD1 Rabbit Polyclonal Antibody
Catalog # BWRLBS60408
Anti-AMD1 Rabbit Polyclonal Antibody
Catalog # BWRLBS60408

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    AMD1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

A synthetic peptide corresponding to residues in Human AMD1.

AMD1 pAb detects endogenous levels of AMD1 protein.

Type: Primary
Antigen: AMD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat