Anti-GALK1 Rabbit Polyclonal Antibody
Catalog # BWRLBS5729
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:GALK1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 4-49 of Human GALK1.
GALK1 pAb detects endogenous levels of GALK1 protein.
Type: Primary
Antigen: GALK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat