Anti-COX5A Rabbit Polyclonal Antibody
Catalog # BWRLBS5674
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:COX5A
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 53-98 of Human COX5A.
COX5A pAb detects endogenous levels of COX5A protein.
Type: Primary
Antigen: COX5A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat