Anti-GEMIN2 Rabbit Polyclonal Antibody
100 µg
Catalog # BWRLBS60255
Supplier: Bioworld Technology
Jump to:
Product Details & Documents
A synthetic peptide corresponding to residues in Human GEMIN2.
GEMIN2 pAb detects endogenous levels of GEMIN2 protein.
Type: Primary
Antigen: GEMIN2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Specifications
- Size:100 µg
- Western blot:Yes
- Cat. No.:BWRLBS60255
- Clonality:Polyclonal
- Reactivity:Human|Rat|Mouse
- Antibody type:Primary
- Host:Rabbit
- Antigen symbol:GEMIN2



