Anti-TOR1A Rabbit Polyclonal Antibody
Catalog # BWRLBS6589
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:TOR1A
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human TOR1A.
TOR1A pAb detects endogenous levels of TOR1A protein.
Type: Primary
Antigen: TOR1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat