Anti-CYP11A1 Rabbit Polyclonal Antibody
Catalog # BWRLBS6578
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CYP11A1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human CYP11A1.
CYP11A1 pAb detects endogenous levels of CYP11A1 protein.
Type: Primary
Antigen: CYP11A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat