Anti-CDK6 Rabbit Polyclonal Antibody
Catalog # BWRLBS6559
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CDK6
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human CDK6.
CDK6 pAb detects endogenous levels of CDK6 protein.
Type: Primary
Antigen: CDK6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat