Anti-AMACR Rabbit Polyclonal Antibody
Catalog # BWRLBS6159
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:AMACR
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human AMACR.
AMACR pAb detects endogenous levels of AMACR protein.
Type: Primary
Antigen: AMACR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human