Anti-GDF-15 Rabbit Polyclonal Antibody
BWRLBS6152
- Antibody type:Primary
- Antigen symbol:GDF-15
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human GDF-15.
GDF15 pAb detects endogenous levels of GDF15 protein.
Type: Primary
Antigen: GDF-15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human