Order Entry
Export
ContactUsLinkComponent
Anti-p-MEK1 Rabbit Polyclonal Antibody [clone: T286]
Anti-p-MEK1 Rabbit Polyclonal Antibody [clone: T286]
Catalog # BWRLBS4844
Anti-p-MEK1 Rabbit Polyclonal Antibody [clone: T286]
Catalog # BWRLBS4844

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-MEK1
  • Clonality:
    Polyclonal
  • Clone:
    T286
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human MEK1 around the phosphorylation site of Threonine 286.

p-MEK1 (T286) pAb detects endogenous levels of MEK1 protein only when phosphorylated at Thr286.

Type: Primary
Antigen: p-MEK1
Clonality: Polyclonal
Clone: T286
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat